missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylbenzenesulfonyl chloride, 98%

Product Code. p-7044659
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311997 1 g 1g
11321997 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311997 Supplier Thermo Scientific Alfa Aesar Supplier No. L07629.03
Add to basket
This item is not returnable. View return policy

CAS: 25300-37-2 | C7H6Cl2O2S | 225.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25300-37-2
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.08
MDL Number MFCD00051767
InChI Key FJVZVTWCAGCLCS-UHFFFAOYSA-N
Synonym 2-chloro-6-methylbenzene-1-sulfonyl chloride, 2-chloro-6-methylbenzenesulfonylchloride, 2-chloro-6-methyl-benzenesulfonyl chloride, acmc-20a0m7, chloro 2-chloro-6-methylphenyl sulfone, 2-chloro-6-methylphenylsulfonyl chloride, 2-chloro-6-methylbenzene sulfonyl chloride, 2-chloro-6-methylbenzene-sulfonyl chloride, 2-chloro-6-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2-chloro-6-methyl
PubChem CID 15725370
SMILES CC1=C(C(Cl)=CC=C1)S(Cl)(=O)=O

Specifications

Boiling Point 132°C to 134°C (0.5 mmHg)
Refractive Index 1.581
Quantity 1 g
UN Number UN3265
Beilstein 2722659
Sensitivity Moisture sensitive
IUPAC Name 2-chloro-6-methylbenzene-1-sulfonyl chloride
Formula Weight 225.09
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.