missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitro-N-(4-pyridyl)benzamide, Thermo Scientific™

Product Code. 15436399
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 15436399

Brand: Thermo Scientific Alfa Aesar J64455.MA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • A cell-permeable chloro-nitro-benzamido compound that acts as a potent, specific, irreversible, and high-affinity antagonist of PPAR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313516-66-4
Molecular Formula C12H8ClN3O3
Molecular Weight (g/mol) 277.664
MDL Number MFCD00121849
InChI Key FRPJSHKMZHWJBE-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-n-4-pyridinylbenzamide, 2-chloro-5-nitro-n-pyridin-4-yl benzamide, 2-chloro-5-nitro-n-4-pyridyl benzamide, benzamide, 2-chloro-5-nitro-n-4-pyridinyl, 2-chloro-5-nitro-n-4-pyridinyl-benzamide, d03bpg, d08gqc, 2-chloro-5-nitro-n-4-pyridinyl benzamide, benzamide,2-chloro-5-nitro-n-4-pyridinyl, n1-4-pyridyl-2-chloro-5-nitrobenzamide
PubChem CID 2777391
IUPAC Name 2-chloro-5-nitro-N-pyridin-4-ylbenzamide
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)NC2=CC=NC=C2)Cl

Specifications

Color White
Quantity 10 mg
Sensitivity Light sensitive
Solubility Information Soluble in DMSO at 10mg/ml. Insoluble in water.
Formula Weight 277.66
Physical Form Solid
Chemical Name or Material 2-Chloro-5-nitro-N-(4-pyridyl)benzamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.