Learn More
(2-Chloro-5-iodophenyl)(4-fluorophenyl)methanone, TRC
Specifications
Specifications
| Chemical Name or Material | (2-Chloro-5-iodophenyl)(4-fluorophenyl)methanone |
| CAS | 915095-86-2 |
| Molecular Formula | C13H7ClFIO |
| Quantity | 2.5 g |
| Synonym | Methanone, (2-chloro-5-iodophenyl)(4-fluorophenyl)- (9CI, ACI), (2-Chloro-5-iodophenyl)(4-fluorophenyl)methanone (ACI) |
| InChI Formula | InChI=1S/C13H7ClFIO/c14-12-6-5-10(16)7-11(12)13(17)8-1-3-9(15)4-2-8/h1-7H |
| SMILES | Fc1ccc(cc1)C(=O)c2cc(I)ccc2Cl |
| IUPAC Name | (2-chloro-5-iodophenyl)-(4-fluorophenyl)methanone |
| Molecular Weight (g/mol) | 360.55 |
| Formula Weight | 359.9214 |