missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoro-3-(trifluoromethyl)benzoic Acid, TRC

Product Code. 30359325
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359325 100 mg 100mg
30356835 50 mg -
30368434 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359325 Supplier Toronto Research Chemicals Supplier No. TRCC386253100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1805955-65-0
Molecular Formula C8H3O2F4Cl
Molecular Weight (g/mol) 242.55
IUPAC Name 2-chloro-5-fluoro-3-(trifluoromethyl)benzoic acid
SMILES OC(=O)c1cc(F)cc(c1Cl)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H3ClF4O2/c9-6-4(7(14)15)1-3(10)2-5(6)8(11,12)13/h1-2H,(H,14,15)
Formula Weight 241.9758
Chemical Name or Material 2-Chloro-5-fluoro-3-(trifluoromethyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.