Learn More
2-Chloro-5,6-dimethyl-3-pyridinecarbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-5,6-dimethyl-3-pyridinecarbonitrile |
| CAS | 65176-93-4 |
| Molecular Formula | C8H7ClN2 |
| Quantity | 500 mg |
| Synonym | 2-Chloro-3-cyano-5,6-dimethylpyridine |
| InChI Formula | InChI=1S/C8H7ClN2/c1-5-3-7(4-10)8(9)11-6(5)2/h3H,1-2H3 |
| SMILES | Cc1cc(C#N)c(Cl)nc1C |
| IUPAC Name | 2-chloro-5,6-dimethylpyridine-3-carbonitrile |
| Molecular Weight (g/mol) | 166.61 |
| Formula Weight | 166.0298 |