Learn More
2-Chloro-5-(1-methyl-2-pyrrolidinyl)pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-5-(1-methyl-2-pyrrolidinyl)pyridine |
| CAS | 185510-30-9 |
| Molecular Formula | C10 H17 N7 O8 S |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C10H17N7O8S/c11-6-15-5-3(2-24-8(13)18)14-7(12)17-1-4(25-26(21,22)23)10(19,20)9(5,17)16-6/h3-5,19-20H,1-2H2,(H2,12,14)(H2,13,18)(H3,11,15,16)(H,21,22,23)/t3?,4-,5-,9+/m1/s1 |
| SMILES | NC(=O)OCC1N=C(N)N2C[C@@H](OS(=O)(=O)O)C(O)(O)[C@@]23NC(=N[C@H]13)N |
| IUPAC Name | [(3aR,9R,10aS)-2,6-diamino-10,10-dihydroxy-9-sulfooxy-3a,4,8,9-tetrahydro-1H-pyrrolo[1,2-c]purin-4-yl]methyl carbamate |
| Molecular Weight (g/mol) | 395.349 |
| Formula Weight | 395.086 |