missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)benzonitrile, 98%, Thermo Scientific™

Product Code. p-7029434
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411440 1 g 1g
11421440 5 g 5g
11431440 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11411440 Supplier Thermo Scientific Alfa Aesar Supplier No. H26894.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1813-33-8
Molecular Formula C8H3ClF3N
Molecular Weight (g/mol) 205.564
MDL Number MFCD00084939
InChI Key GEHMLBFNZKJDQM-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzonitrile, 3-chloro-4-cyanobenzotrifluoride, 3-chloro-4-cyano-benzotrifluoride, 4-trifluoromethyl-2-chlorobenzonitrile, benzonitrile, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoromethyl benzenecarbonitrile, pubchem4745, acmc-209eho, buttpark 9757-31, ksc494o6f
PubChem CID 2736476
IUPAC Name 2-chloro-4-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)C#N

Specifications

Melting Point 82°C to 83°C (8mmHg)
Density 1.389
Boiling Point 192°C to 193°C
Flash Point >104°C (220°F)
Odor Characteristic
Refractive Index 1.484
Quantity 1 g
UN Number UN3276
Formula Weight 205.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.