Learn More
2-Chloro-4-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™
Product Code. 15424427
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424427 250 mg 250mg
15414427 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15424427 Supplier Thermo Scientific Alfa Aesar Supplier No. H31789.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23228-45-7
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.56
MDL Number MFCD06208178
InChI Key DTIJZEUKFYGSEC-UHFFFAOYSA-N
PubChem CID 11118363
IUPAC Name 2-chloro-4-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=C(Cl)C=C(C=C1)C(F)(F)F
Specifications
Melting Point 114°C to 117°C
Quantity 250 mg
Synonym 2-chloro-4-trifluoromethyl benzoic acid, benzoic acid, 2-chloro-4-trifluoromethyl, pubchem4592, 4-carboxy-3-chlorobenzotrifluoride, 2-chloro-4 trifluoromethyl benzoic acid, 2-chloro-4-trifluoromethyl-benzoic acid, 2-chloro-4-trifluoromethyl benzoicacid, benzoic acid,2-chloro-4-trifluoromethyl
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzoic acid