missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methoxypyridine, 97%

Product Code. 11308646
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11308646

Brand: Thermo Scientific Alfa Aesar L17696.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 52605-96-6 | C6H6ClNO | 143.57 g/mol

It is a pharmaceutical raw material and also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical raw material and also used in medicine.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52605-96-6
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD03426022
InChI Key CCVNZKGBNUPYPG-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-3-methoxy, 2-chloro-3-methoxy-pyridine, pubchem6146, acmc-1ap1j, 2chloro-3-methoxylpyridine, ksc498a8t
PubChem CID 104252
IUPAC Name 2-chloro-3-methoxypyridine
SMILES COC1=C(N=CC=C1)Cl

Specifications

Melting Point 46°C to 48°C
Quantity 1 g
Beilstein 115568
Solubility Information Insoluble in water.
Formula Weight 143.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-methoxypyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.