missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluorobenzaldehyde, 95%, Thermo Scientific™

Product Code. 15407177
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407177 1 g 1g
15417177 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407177 Supplier Thermo Scientific Alfa Aesar Supplier No. H34238.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96516-31-3
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD07782053
InChI Key PIZVRLVKXWEMGO-UHFFFAOYSA-N
Synonym 2-chloro-3-fluoro-benzaldehyde, benzaldehyde, 2-chloro-3-fluoro, pubchem1432, 2-chlorofluorobenzaldehyde, ksc486i8l
PubChem CID 14944905
IUPAC Name 2-chloro-3-fluorobenzaldehyde
SMILES FC1=CC=CC(C=O)=C1Cl

Specifications

Melting Point 29°C to 32°C
Color White
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 158.56
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-fluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.