missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylhexane, 97%

Product Code. p-7024918
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479616 5 g 5g
11489616 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479616 Supplier Thermo Scientific Alfa Aesar Supplier No. B20183.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4398-65-6 | C7H15Cl | 134.65 g/mol

2-Chloro-2-methylhexane is used to produce other chemicals. For example, it is used to produce (1, 1-Dimethyl-pentyl)-benzene. Also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-2-methylhexane is used to produce other chemicals. For example, it is used to produce (1, 1-Dimethyl-pentyl)-benzene. Also used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4398-65-6
Molecular Formula C7H15Cl
Molecular Weight (g/mol) 134.65
MDL Number MFCD00060779
InChI Key KBOBQLJBYKKAPN-UHFFFAOYSA-N
Synonym hexane, 2-chloro-2-methyl, acmc-1an7f, 2-chloro-2-methyl-hexane, 1,1-dimethylpentyl chloride, 1-chloro-1,1-dimethylpentane, 2-chloro-2-methylhexane 5g
PubChem CID 138224
IUPAC Name 2-chloro-2-methylhexane
SMILES CCCCC(C)(C)Cl

Specifications

Quantity 5 g
UN Number UN1993
Formula Weight 134.65
Percent Purity 97%
Chemical Name or Material 2-Chloro-2-methylhexane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.