Learn More
2-Chloro-10-(3-chloropropyl)phenothiazine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-10-(3-chloropropyl)phenothiazine |
| CAS | 2765-59-5 |
| Molecular Formula | C15H13Cl2NS |
| Quantity | 25 mg |
| Synonym | 2-Chloro-10-(3-chloropropyl)-10H-phenothiazine, gamma-(2-Chlorophenothiazinyl-10)propyl Chloride |
| InChI Formula | InChI=1S/C15H13Cl2NS/c16-8-3-9-18-12-4-1-2-5-14(12)19-15-7-6-11(17)10-13(15)18/h1-2,4-7,10H,3,8-9H2 |
| SMILES | ClCCCN1c2ccccc2Sc3ccc(Cl)cc13 |
| IUPAC Name | 2-chloro-10-(3-chloropropyl)phenothiazine |
| Molecular Weight (g/mol) | 310.24 |
| Formula Weight | 309.0146 |