Learn More
2-Chloro-1-(4-hydroxy-3-methoxyphenyl)propan-1-one, TRC
Product Code. 30351855
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30351855 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30351855 Supplier Toronto Research Chemicals Supplier No. TRCC368480100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-1-(4-hydroxy-3-methoxyphenyl)propan-1-one
CAS 68505-86-2
Molecular Formula C10H11ClO3
Quantity 100 mg
Synonym 2-Chloro-4'-hydroxy-3'-methoxypropiophenone
InChI Formula InChI=1S/C10H11ClO3/c1-6(11)10(13)7-3-4-8(12)9(5-7)14-2/h3-6,12H,1-2H3
SMILES O=C(C1=CC=C(O)C(OC)=C1)C(Cl)C
IUPAC Name 2-chloro-1-(4-hydroxy-3-methoxyphenyl)propan-1-one
Molecular Weight (g/mol) 214.65
Formula Weight 214.04