Learn More
2-Butyloctanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Butyloctanoic Acid |
| CAS | 27610-92-0 |
| Molecular Formula | C12H24O2 |
| Quantity | 250 mg |
| Synonym | Isocarb 12, Jaric I 12, alpha-Butylcaprylic Acid, 2-Butyl-caprylic acid |
| InChI Formula | InChI=1S/C12H24O2/c1-3-5-7-8-10-11(12(13)14)9-6-4-2/h11H,3-10H2,1-2H3,(H,13,14) |
| SMILES | CCCCCCC(CCCC)C(=O)O |
| IUPAC Name | 2-butyloctanoic acid |
| Molecular Weight (g/mol) | 200.32 |
| Formula Weight | 200.1776 |