Learn More
2-Butyl-4,6-dihydroxybenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Butyl-4,6-dihydroxybenzoic acid |
| CAS | 173324-41-9 |
| Molecular Formula | C11 H14 O4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C11H14O4/c1-2-3-4-7-5-8(12)6-9(13)10(7)11(14)15/h5-6,12-13H,2-4H2,1H3,(H,14,15) |
| SMILES | CCCCc1cc(O)cc(O)c1C(=O)O |
| IUPAC Name | 2-butyl-4,6-dihydroxy-benzoic acid |
| Molecular Weight (g/mol) | 210.23 |
| Formula Weight | 210.0892 |