missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine, TRC

Product Code. 30006809
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006809 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30006809 Supplier Toronto Research Chemicals Supplier No. TRCB68672525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-04-6
Formula Weight 156.9527
InChI Formula InChI=1S/C5H4BrN/c6-5-3-1-2-4-7-5/h1-4H
IUPAC Name 2-bromopyridine
Molecular Formula C5H4BrN
Molecular Weight (g/mol) 158
SMILES Brc1ccccn1
Synonym 2-Bromopyridin, 2-Pyridyl Bromide, NSC 8031, Pyridin-2-yl bromide, o-Bromopyridine, beta-Bromopyridine;
Chemical Name or Material 2-Bromopyridine

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.