missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomethyl-4-fluorobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-7054716
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
421552-12-7
C8H5BrFN
214.04
MFCD08059542
CHCAGFNTASDQFX-UHFFFAOYSA-N
2-cyano-5-fluorobenzyl bromide, 2-bromomethyl-4-fluorobenzonitrile, 2-cyano-5-fluorobenzylbromide, 2-cyano-5-fluoro benzyl bromide, 2-bromomethyl-4-fluoro-benzonitrile, 2-bromomethyl-4-fluorobenzenecarbonitrile, pubchem4906, acmc-1arut, ksc493s9t, 2-cyano-5-flurobenzyl bromide
21984990
2-(bromomethyl)-4-fluorobenzonitrile
FC1=CC(CBr)=C(C=C1)C#N
This item is not returnable. View return policy
421552-12-7
C8H5BrFN
214.04
MFCD08059542
CHCAGFNTASDQFX-UHFFFAOYSA-N
2-cyano-5-fluorobenzyl bromide, 2-bromomethyl-4-fluorobenzonitrile, 2-cyano-5-fluorobenzylbromide, 2-cyano-5-fluoro benzyl bromide, 2-bromomethyl-4-fluoro-benzonitrile, 2-bromomethyl-4-fluorobenzenecarbonitrile, pubchem4906, acmc-1arut, ksc493s9t, 2-cyano-5-flurobenzyl bromide
21984990
2-(bromomethyl)-4-fluorobenzonitrile
FC1=CC(CBr)=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2923
Formula Weight 214.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromomethyl-4-fluorobenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'