missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzothiazole, 99%

Product Code. p-7054861
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449908 1 g 1g
15459908 5 g 5g
15469908 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15449908 Supplier Thermo Scientific Alfa Aesar Supplier No. H63286.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2516-40-7 | C7H4BrNS | 214.08 g/mol

2-Bromobenzothiazole, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzothiazole, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-40-7
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD02681887
InChI Key DRLMMVPCYXFPEP-UHFFFAOYSA-N
Synonym 2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou
PubChem CID 612040
IUPAC Name 2-bromo-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)Br

Specifications

Melting Point 39°C to 43°C
Color Yellow
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 214.08
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2-Bromobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.