Learn More
2-Bromoadenine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromoadenine |
| CAS | 28128-25-8 |
| Molecular Formula | C5 H4 Br N5 |
| Quantity | 100 mg |
| Synonym | 2-Bromo-9H-purin-6-amine, 2-Bromo-1H-purin-6-amine, 2-Bromo-7H-purin-6-amine, 2-Bromoadenine, NSC 52757 |
| InChI Formula | InChI=1S/C5H4BrN5/c6-5-10-3(7)2-4(11-5)9-1-8-2/h1H,(H3,7,8,9,10,11) |
| SMILES | Nc1nc(Br)nc2nc[nH]c12 |
| IUPAC Name | 2-bromo-7H-purin-6-amine |
| Molecular Weight (g/mol) | 214.023 |
| Formula Weight | 212.965 |