Learn More
2-Bromo-6-chloroaniline, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-6-chloroaniline |
| CAS | 59772-49-5 |
| Molecular Formula | C6 H5 Br Cl N |
| Quantity | 1 g |
| Synonym | 2-Bromo-6-chloroaniline, 2-Bromo-6-chlorobenzenamine |
| InChI Formula | InChI=1S/C6H5BrClN/c7-4-2-1-3-5(8)6(4)9/h1-3H,9H2 |
| SMILES | Nc1c(Cl)cccc1Br |
| IUPAC Name | 2-bromo-6-chloroaniline |
| Molecular Weight (g/mol) | 206.47 |
| Formula Weight | 204.9294 |