Learn More
2-Bromo-5-fluorobenzoic acid, 98+%
Product Code. 11457076
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457076 5 g 5g
11447076 1 g 1g
11349669 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11457076 Supplier Thermo Scientific Alfa Aesar Supplier No. B25394.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 394-28-5 | C7H3BrFO2 | 218.00 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 394-28-5
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD00142874
InChI Key OQBMJMJZMDBQSM-UHFFFAOYSA-M
Synonym benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747
PubChem CID 2778181
IUPAC Name 2-bromo-5-fluorobenzoic acid
SMILES [O-]C(=O)C1=CC(F)=CC=C1Br
Specifications
Melting Point 154°C to 158°C
Quantity 5 g
Beilstein 2575978
Formula Weight 219.01
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-fluorobenzoic acid