missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorobenzaldehyde, 98%

Product Code. 15411909
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411909 250 mg 250mg
15491899 1 g 1g
15401909 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411909 Supplier Thermo Scientific Alfa Aesar Supplier No. H64621.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 476620-54-9 | C7H3BrF2O | 221.001 g/mol

It is an important intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 476620-54-9
Molecular Formula C7H3BrF2O
Molecular Weight (g/mol) 221.001
MDL Number MFCD16659662
InChI Key ZHSZEWFWVDLMDL-UHFFFAOYSA-N
Synonym benzaldehyde, 2-bromo-4,5-difluoro, acmc-209ka1, 2-bromo-4,5-difluoro-benzaldehyde
PubChem CID 21867425
IUPAC Name 2-bromo-4,5-difluorobenzaldehyde
SMILES C1=C(C(=CC(=C1F)F)Br)C=O

Specifications

Color White
Quantity 250 mg
Beilstein 20365202
Solubility Information Slightly soluble in water.
Formula Weight 221
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Bromo-4,5-difluorobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.