missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoropropionic acid, 96%, Thermo Scientific Chemicals

Product Code. 15494337
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494337 250 mg 250mg
15484337 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494337 Supplier Thermo Scientific Alfa Aesar Supplier No. H31718.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16652-36-1 | Molecular Formula: C3H4BrFO2 | Molecular Weight: 170.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16652-36-1
Molecular Formula C3H4BrFO2
Molecular Weight (g/mol) 170.97
MDL Number MFCD09800640
InChI Key FBPUCVAAEPPEMG-UHFFFAOYNA-N
Synonym 2-bromo-3-fluoropropionic acid, acmc-20ao2z, 3-fluoro-2-bromo-propionic acid
PubChem CID 316511
IUPAC Name 2-bromo-3-fluoropropanoic acid
SMILES OC(=O)C(Br)CF

Specifications

Boiling Point 62°C to 63°C (0.2 mmHg)
Refractive Index 1.4655
Quantity 250 mg
UN Number UN3265
Formula Weight 170.97
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-fluoropropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.