Learn More
2-Bromo-1,6-dimethyl Ester Hexanedioic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-1,6-dimethyl Ester Hexanedioic Acid |
| CAS | 3196-19-8 |
| Molecular Formula | C16 H18 N O10 S . Na |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C16H19NO10S.Na/c1-7-4-12(19)26-10-5-8(2-3-9(7)10)25-16-13(17-28(22,23)24)15(21)14(20)11(6-18)27-16;/h2-5,11,13-18,20-21H,6H2,1H3,(H,22,23,24);/q;+1/p-1/t11?,13?,14-,15?,16+;/m0./s1 |
| SMILES | [Na+].CC1=CC(=O)Oc2cc(O[C@@H]3OC(CO)[C@H](O)C(O)C3NS(=O)(=O)[O-])ccc12 |
| IUPAC Name | sodium;N-[(2S,5R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]sulfamate |
| Molecular Weight (g/mol) | 439.37 |
| Formula Weight | 439.055 |