Learn More
2-Benzylcyclohexanone, 97%
Product Code. 11397585
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397585 1 g 1g
11307595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11397585 Supplier Thermo Scientific Alfa Aesar Supplier No. L05776.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 946-33-8 | C13H16O | 188.27 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 946-33-8
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00051455
InChI Key CUYLPYBYCYQGCE-UHFFFAOYSA-N
Synonym 2-benzylcyclohexanone, cyclohexanone, 2-phenylmethyl, 2-benzyl cyclohexanone, 2-benzyl-cyclohexan-1-one, 2-phenylmethyl cyclohexanone, cyclohexanone,2-phenylmethyl, 2rs-2-phenylmethylcyclohexanone
PubChem CID 98945
IUPAC Name 2-benzylcyclohexan-1-one
SMILES C1CCC(=O)C(C1)CC2=CC=CC=C2
Specifications
Melting Point 28°C to 30°C
Density 1.03
Boiling Point 103°C to 105°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 1 g
Beilstein 2046670
Formula Weight 188.27
Percent Purity 97%
Chemical Name or Material 2-Benzylcyclohexanone