Learn More
2-Benzylamino-2-methyl-1-propanol, TRC
Spezifikation
Spezifikation
| Chemical Name or Material | 2-Benzylamino-2-methyl-1-propanol |
| CAS | 10250-27-8 |
| Molecular Formula | C11 H17 N O |
| Quantity | 500 mg |
| Synonym | 1-Propanol, 2-methyl-2-[(phenylmethyl)amino]-, 1-Propanol, 2-(benzylamino)-2-methyl- (7CI,8CI), 2-Methyl-2-[(phenylmethyl)amino]-1-propanol, (Benzyl)(2-hydroxy-1,1-dimethylethyl)amine, 2-(Benzylamino)-2-methyl-1-propanol, 2-(Benzylamino)-2-methyl-propanol |
| InChI Formula | InChI=1S/C11H17NO/c1-11(2,9-13)12-8-10-6-4-3-5-7-10/h3-7,12-13H,8-9H2,1-2H3 |
| SMILES | CC(C)(CO)NCc1ccccc1 |
| IUPAC Name | 2-(benzylamino)-2-methylpropan-1-ol |
| Molecular Weight (g/mol) | 179.26 |
| Formula Weight | 179.131 |