Learn More
2-Benzamido-3-methylbutanoic acid, 95%, BLD Pharm™
Product Code. 30968377
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30968377 10 g 10g
30954627 25 g 25g
30963569 5 g 5g
30953140 1 g 1g
30952794 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30968377 Supplier BLD Pharm Supplier No. BD11595810g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 2901-80-6
Molecular Formula C12H15NO3
MDL Number MFCD00038283
Quantity 10 g
SMILES CC(C)C(C(O)=O)NC(C1=CC=CC=C1)=O
Molecular Weight (g/mol) 221.25
Percent Purity 95%
Chemical Name or Material 2-Benzamido-3-methylbutanoic acid