Learn More
2-Azabicyclo[2.2.1]heptane, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Azabicyclo[2.2.1]heptane |
| CAS | 279-24-3 |
| Molecular Formula | C6H11N |
| Quantity | 250 mg |
| Synonym | 2-Azanorbornane, 3-Azabicyclo[2.2.1]heptane |
| InChI Formula | InChI=1S/C6H11N/c1-2-6-3-5(1)4-7-6/h5-7H,1-4H2 |
| SMILES | C1CC2CC1CN2 |
| IUPAC Name | 2-Azabicyclo[2.2.1]heptane |
| Molecular Weight (g/mol) | 97.16 |
| Formula Weight | 97.09 |