missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine-3-carboxaldehyde, 98%

Product Code. p-7047679
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11355395 1 g 1g
11365395 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11355395

Brand: Thermo Scientific Alfa Aesar L19620.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7521-41-7 | C6H6N2O | 122.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7521-41-7
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD01830382
InChI Key NXMFJCRMSDRXLD-UHFFFAOYSA-N
Synonym 2-aminonicotinaldehyde, 2-amino-3-pyridinecarboxaldehyde, 2-amino-3-formylpyridine, 3-pyridinecarboxaldehyde, 2-amino, 2-amino-pyridine-3-carbaldehyde, 2-aminopyridine-3-carboxaldehyde, 2-amino-3-pyridine carboxaldehyde, 2-amino-pyridin-3-carbaldehyde, pubchem1262, 2-amino nicotinaldehyde
PubChem CID 737633
IUPAC Name 2-aminopyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)N)C=O

Specifications

Melting Point 99°C to 102°C
Quantity 1 g
Beilstein 109598
Sensitivity Air sensitive
Formula Weight 122.13
Percent Purity 98%
Chemical Name or Material 2-Aminopyridine-3-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.