Learn More
2-Aminoindole Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Aminoindole, Hydrochloride |
| CAS | 27878-37-1 |
| Molecular Formula | C8 H8 N2 . Cl H |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C8H8N2.ClH/c9-8-5-6-3-1-2-4-7(6)10-8;/h1-5,10H,9H2;1H |
| SMILES | Cl.Nc1cc2ccccc2[nH]1 |
| IUPAC Name | 1H-indol-2-amine;hydrochloride |
| Molecular Weight (g/mol) | 168.62 |
| Formula Weight | 168.0454 |