Learn More
2-Aminoindan-2-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Aminoindan-2-carboxylic Acid |
| CAS | 27473-62-7 |
| Molecular Formula | C10H11NO2 |
| Quantity | 250 mg |
| Synonym | 2-Amino-2,3-dihydro-1H-indene-2-carboxylic Acid, H-Aic-OH |
| InChI Formula | InChI=1S/C10H11NO2/c11-10(9(12)13)5-7-3-1-2-4-8(7)6-10/h1-4H,5-6,11H2,(H,12,13) |
| SMILES | OC(C1(CC(C=CC=C2)=C2C1)N)=O |
| IUPAC Name | 2-amino-1,3-dihydroindene-2-carboxylic acid |
| Molecular Weight (g/mol) | 177.2 |
| Formula Weight | 177.079 |