Learn More
2-Amino Nevirapine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino Nevirapine |
| CAS | 284686-15-3 |
| Molecular Formula | C15H15N5O |
| Quantity | 10 mg |
| Synonym | 2-Amino-11-cyclopropyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one |
| InChI Formula | InChI=1S/C15H15N5O/c1-8-7-11(16)18-14-12(8)19-15(21)10-3-2-6-17-13(10)20(14)9-4-5-9/h2-3,6-7,9H,4-5H2,1H3,(H2,16,18)(H,19,21) |
| SMILES | CC1=CC(=N)NC2=C1N=C(O)c3cccnc3N2C4CC4 |
| IUPAC Name | 5-amino-2-cyclopropyl-7-methyl-2,4,9,15-tetrazatricyclo[9.4.0.03,8]pentadeca-1(11),3(8),4,6,12,14-hexaen-10-one |
| Molecular Weight (g/mol) | 281.31 |
| Formula Weight | 281.1277 |