Learn More
2-amino-N-(quinolin-8-yl)acetamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-amino-N-(quinolin-8-yl)acetamide |
| CAS | 587832-36-8 |
| Molecular Formula | C11H11N3O |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C11H11N3O/c12-7-10(15)14-9-5-1-3-8-4-2-6-13-11(8)9/h1-6H,7,12H2,(H,14,15) |
| SMILES | NCC(=O)Nc1cccc2cccnc12 |
| IUPAC Name | 2-amino-N-quinolin-8-ylacetamide |
| Molecular Weight (g/mol) | 201.22 |
| Formula Weight | 201.0902 |