Learn More
2-Amino-acetonitrile-2,2-d2 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-acetonitrile-2,2-d2 Hydrochloride |
| CAS | 1221897-87-5 |
| Molecular Formula | C2H2D2N2 . HCl |
| Quantity | 250 mg |
| Synonym | 2-Amino-acetonitrile-d2 Hydrochloride, Amino-acetonitrile-d2 Monohydrochloride, Glycinonitrile-d2 Monohydrochloride, 2-Aminoacetonitrile-d2 Hydrochloride, Aminoacetonitrile-d2 Hydrochloride, Aminoacetonitrile-d2 Monohydrochloride, Glycinonitrile-d2 Hydrochloride, α-Aminoacetonitrile-d2 Hydrochloride |
| InChI Formula | InChI=1S/C2H4N2.ClH/c3-1-2-4;/h1,3H2;1H/i1D2; |
| SMILES | NC([2H])([2H])C#N.Cl |
| IUPAC Name | 2-aminoacetonitrile-d2 hydrogen chloride |
| Molecular Weight (g/mol) | 94.54 |
| Formula Weight | 94.03 |