Learn More
2-Amino-5-chlorobenzenesulfonamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-5-chlorobenzenesulfonamide |
| CAS | 5790-69-2 |
| Molecular Formula | C6H7ClN2O2S |
| Quantity | 1 g |
| Synonym | Hydrochlorothiazide Impurity 28 |
| InChI Formula | InChI=1S/C6H7ClN2O2S/c7-4-1-2-5(8)6(3-4)12(9,10)11/h1-3H,8H2,(H2,9,10,11) |
| SMILES | Nc1ccc(Cl)cc1S(=O)(=O)N |
| IUPAC Name | 2-amino-5-chlorobenzenesulfonamide |
| Molecular Weight (g/mol) | 206.65 |
| Formula Weight | 205.9917 |