Learn More
2-Amino-5-bromo-3-methylpyridine, TRC
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-5-bromo-3-methylpyridine |
| CAS | 3430-21-5 |
| Molecular Formula | C6 H7 Br N2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C6H7BrN2/c1-4-2-5(7)3-9-6(4)8/h2-3H,1H3,(H2,8,9) |
| SMILES | Cc1cc(Br)cnc1N |
| IUPAC Name | 5-bromo-3-methylpyridin-2-amine |
| Molecular Weight (g/mol) | 187.04 |
| Formula Weight | 185.9793 |