missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile

Product Code. p-7031103
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487975 1 g 1g
15497975 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487975 Supplier Thermo Scientific Alfa Aesar Supplier No. H32059.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 70291-62-2 | C8H8N2S | 164.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70291-62-2
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00121495
InChI Key RILAXFOADRDGPQ-UHFFFAOYSA-N
PubChem CID 258182
IUPAC Name 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
SMILES NC1=C(C#N)C2=C(CCC2)S1

Specifications

Melting Point 153°C to 154°C
Quantity 1 g
UN Number UN3439
Synonym 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carbonitrile, 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carbonitrile, 2-amino-4,5-trimethylene-3-thiophenecarbonitrile, 4h-cyclopenta b thiophene-3-carbonitrile, 2-amino-5,6-dihydro, 2-amino-3-cyano-5,6-dihydro-4h-cyclopenta b thiophene, 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carbonitrile, 4h-cyclopenta b thiophene-3-carbonitrile,2-amino-5,6-dihydro, maybridge1_003206, 4,5-trimethylene-2-aminothiophene-3-carbonitrile, 2-amino-5,6-dihydro-4h-cyclopenta b thioph ene-3-carbonitrile
Formula Weight 164.23
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.