Learn More
2-Amino-3-cyano-5-iodopyridine, 97%
Product Code. 15427127
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15427127
Brand: Thermo Scientific Alfa Aesar H34191.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 1347815-41-1 | C6H4IN3 | 245.023 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1347815-41-1
Molecular Formula C6H4IN3
Molecular Weight (g/mol) 245.023
MDL Number MFCD20040120
InChI Key NJLCSLOABAUGHW-UHFFFAOYSA-N
Synonym 2-amino-5-iodonicotinonitrile, 2-amino-3-cyano-5-iodopyridine, 2-amino-5-iodo-nicotinonitrile
PubChem CID 65815340
IUPAC Name 2-amino-5-iodopyridine-3-carbonitrile
SMILES C1=C(C(=NC=C1I)N)C#N
Specifications
Melting Point 193°C to 197°C
Quantity 5 g
UN Number UN3439
Formula Weight 245.02
Percent Purity 97%
Chemical Name or Material 2-Amino-3-cyano-5-iodopyridine