Learn More
2-Amino-3-chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Product Code. 10696250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10696250 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10696250 Supplier Thermo Scientific Acros Supplier No. 436270250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
CAS 79456-26-1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 95°C to 97.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H4ClF3N2
MDL Number MFCD00042154
Quantity 25 g
Synonym 3-chloro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-chloro-5-trifluoromethyl pyridine, 2-amino-3-chloro-5-trifluoromethylpyridine, 3-chloro-5-trifluoromethyl-2-pyridinamine, 3-chloro-5-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridin-2-ylamine, 2-amino-3-chloro-5-trifluoropyridine, pubchem2974
InChI Key WXNPZQIRDCDLJD-UHFFFAOYSA-N
SMILES NC1=C(Cl)C=C(C=N1)C(F)(F)F
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol) 196.56
PubChem CID 607248
Formula Weight 196.56
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-chloro-5-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-chloro-5-(trifluoromethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.