Learn More
2-(Allylthio)nicotinic acid, 98%
Product Code. 15428869
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428869 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15428869 Supplier Thermo Scientific Alfa Aesar Supplier No. L07090.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 175135-25-8 | C9H9NO2S | 195.24 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 175135-25-8
Melting Point 144°C to 149°C
Molecular Formula C9H9NO2S
MDL Number MFCD00051681
Quantity 5 g
Synonym 2-allylthio nicotinic acid, 2-prop-2-en-1-ylsulfanyl pyridine-3-carboxylic acid, 2-prop-2-enylthiopyridine-3-carboxylic acid, acmc-20aooc, maybridge1_001317, 3-pyridinecarboxylic acid, 2-2-propenylthio, 3-pyridinecarboxylicacid, 2-2-propen-1-ylthio
InChI Key KSBMXXMQUGZNSF-UHFFFAOYSA-N
SMILES OC(=O)C1=C(SCC=C)N=CC=C1
IUPAC Name 2-(prop-2-en-1-ylsulfanyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol) 195.24
PubChem CID 1810568
Formula Weight 195.24
Percent Purity 98%
Chemical Name or Material 2-(Allylthio)nicotinic acid