Learn More
2-Allyl-2-phenyl-4-pentenenitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Allyl-2-phenyl-4-pentenenitrile |
| CAS | 28049-67-4 |
| Molecular Formula | C14 H15 N |
| Quantity | 10 g |
| Synonym | alpha,alpha-Di-2-propen-1-ylbenzeneacetonitrile, Diallylphenylacetonitrile |
| InChI Formula | InChI=1S/C14H15N/c1-3-10-14(12-15,11-4-2)13-8-6-5-7-9-13/h3-9H,1-2,10-11H2 |
| SMILES | C=CCC(CC=C)(C#N)c1ccccc1 |
| IUPAC Name | 2-phenyl-2-prop-2-enylpent-4-enenitrile |
| Molecular Weight (g/mol) | 197.276 |
| Formula Weight | 197.12 |