Learn More
2-Acetyl Phenothiazine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Acetyl Phenothiazine |
| CAS | 6631-94-3 |
| Molecular Formula | C14H11NOS |
| Quantity | 25 g |
| Synonym | ML 171, 1-(10H-Phenothiazin-2-yl)ethanone, Methyl Phenothiazin-2-yl Ketone, 2-Acetyl-10H-phenothiazine, 2-Acetylphenothiazine, NSC 169669, NSC 57951 |
| SMILES | CC(=O)c1ccc2Sc3ccccc3Nc2c1 |
| Molecular Weight (g/mol) | 241.31 |