Learn More
2-Acetamido-3-acetoxybenzoic Acid Methyl Ester, TRC
Product Code. 30320881
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320881 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30320881 Supplier Toronto Research Chemicals Supplier No. TRCA1680955MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Acetamido-3-acetoxybenzoic Acid Methyl Ester
CAS 100193-10-0
Molecular Formula C12H13NO5
Quantity 5 mg
Synonym N-Acetyl-3-hydroxyanthranilic Acid Methyl Ester Acetate, 2-(Acetylamino)-3-(acetyloxy)benzoic Acid Methyl Ester
InChI Formula InChI=1S/C12H13NO5/c1-7(14)13-11-9(12(16)17-3)5-4-6-10(11)18-8(2)15/h4-6H,1-3H3,(H,13,14)
SMILES COC(=O)c1cccc(OC(=O)C)c1NC(=O)C
IUPAC Name methyl 2-acetamido-3-acetyloxybenzoate
Molecular Weight (g/mol) 251.24
Formula Weight 251.0794