Learn More
2-Acetamido-1-N-(beta-L-aspartyl)-2-deoxy-beta-D-glucopyranosylamine, TRC
Specifications
Specifications
| Chemical Name or Material | N-[2-(Acetylamino)-2-deoxy-beta-D-glucopyranosyl]-L-asparagine |
| CAS | 2776-93-4 |
| Molecular Formula | C12 H21 N3 O8 |
| Quantity | 10 mg |
| Synonym | N-[2-(Acetylamino)-2-deoxy-β-D-glucopyranosyl]-L-asparagine, Asparagine, N-(2-acetamido-2-deoxy-β-D-glucopyranosyl)- (7CI), Asparagine, N-(2-acetamido-2-deoxy-β-D-glucopyranosyl)-, L- (8CI), N-[2-(Acetylamino)-2-deoxy-β-D-glucopyranosyl]-L-asparagine, 1-L-β-Aspartamido-2-acetamido-1,2-dideoxy-β-D-glucose, 1-N-β-L-Aspartyl-2-acetamido-2-deoxy-β-D-glucopyranosylamine, 2-Acetamido-1-(L-β-aspartamido)-1,2-dideoxy-β-D-glucopyranose, 2-Acetamido-1-(L-β-aspartamido)-1,2-dideoxy-β-D-glucose, 2-Acetamido-1-(β-L-aspartamido)-1,2-dideoxy-β-D-glucose, 2-Acetamido-1-(β'-L-aspartamido)-1,2-dideoxy-β-D-glucose, 2-Acetamido-1-N-(4-L-aspartyl)-2-deoxy-β-D-glucopyranosylamine, 2-Acetamido-1-N-(L-β-aspartyl)-2-deoxy-β-D-glucopyranosylamine, 2-Acetamido-1-β-(L-β-aspartamido)-1,2-dideoxy-D-glucose, 2-Acetamido-N-(L-aspart-4'-oyl)-2-deoxy-β-D-glucopyranosylamine, 4-N-2-Acetamido-2-deoxy-β-D-glucopyranosyl-L-asparagine, N-(4'-Aspartyl)-2-acetamido-2-deoxy-β-D-glucopyranosylamine |
| InChI Formula | InChI=1S/C12H21N3O8/c1-4(17)14-8-10(20)9(19)6(3-16)23-11(8)15-7(18)2-5(13)12(21)22/h5-6,8-11,16,19-20H,2-3,13H2,1H3,(H,14,17)(H,15,18)(H,21,22)/t5-,6+,8+,9+,10+,11+/m0/s1 |
| SMILES | CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1NC(=O)C[C@H](N)C(=O)O |
| IUPAC Name | (2S)-4-[[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]amino]-2-amino-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 335.31 |
| Formula Weight | 335.1329 |