Learn More
(2,6-Dimethylphenoxy)acetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2,6-Dimethylphenoxy)acetic Acid |
| CAS | 13335-71-2 |
| Molecular Formula | C10 H12 O3 |
| Quantity | 5 g |
| Synonym | Acetic acid, 2-(2,6-dimethylphenoxy)-, Acetic acid, (2,6-dimethylphenoxy)- (9CI), Acetic acid, (2,6-xylyloxy)- (6CI,8CI), 2-(2,6-Dimethylphenoxy)acetic acid, (2,6-Dimethylphenoxy)acetic acid, NSC 408599 |
| InChI Formula | InChI=1S/C10H12O3/c1-7-4-3-5-8(2)10(7)13-6-9(11)12/h3-5H,6H2,1-2H3,(H,11,12) |
| SMILES | Cc1cccc(C)c1OCC(=O)O |
| IUPAC Name | 2-(2,6-dimethylphenoxy)acetic acid |
| Molecular Weight (g/mol) | 180.2 |
| Formula Weight | 180.0786 |