Learn More
2,6-Dimethylphenol-3,4,5-d3,OD, CDN
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethylphenol ring,hydroxy-d4 |
| CAS | 285132-85-6 |
| Molecular Formula | C8 2H4 H6 O |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C8H10O/c1-6-4-3-5-7(2)8(6)9/h3-5,9 H,1-2H3/i3D,4 D,5 D/hD |
| SMILES | [2 H]Oc1c(C)c([2 H])c([2 H])c([2 H])c1C |
| IUPAC Name | 1,2,3-trideuterio-5-deuteriooxy-4,6-dimethylbenzene |
| Molecular Weight (g/mol) | 126.19 |
| Formula Weight | 126.0983 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |