Learn More
2,6-Dimethyl-1,4-dioxane (>85%), TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethyl-1,4-dioxane |
| CAS | 10138-17-7 |
| Molecular Formula | C6 H12 O2 |
| Quantity | 25 mg |
| Synonym | 2,6-Dimethyl-1,4-Dioxane |
| InChI Formula | InChI=1S/C6H12O2/c1-5-3-7-4-6(2)8-5/h5-6H,3-4H2,1-2H3 |
| SMILES | CC1COCC(C)O1 |
| IUPAC Name | 2,6-dimethyl-1,4-dioxane |
| Molecular Weight (g/mol) | 116.158 |
| Formula Weight | 116.084 |