missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethoxytoluene, 98%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 37°C to 41°C |
| Boiling Point | 222°C |
| Flash Point | 96°C (204°F) |
| Odor | Aromatic |
| Molecular Formula | C4H7ClMg |
| MDL Number | MFCD00674003 |
| Quantity | 25 g |
| Beilstein | 2045327 |
| Synonym | methallylmagnesium chloride, 2-methylallylmagnesium chloride, magnesium, chloro 2-methyl-2-propenyl, methallyl magnesium chloride, methallyl-magnesium chloride, 2-methylallyl magnesium chloride, bjvfgwybolmuem-uhfffaoysa-m, 2-methylallylmagnesium chloride solution, 2-methyl-2-propenyl magnesium chloride, 2-methyl-2-propen-1-yl magnesium chloride |
| Solubility Information | Insoluble in water. |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction