Learn More
2,6-Diethylaniline-d15, CDN
Specifications
Specifications
| Chemical Name or Material | 2,6-Diethylaniline-D15 |
| CAS | 285132-89-0 |
| Molecular Formula | C10 D15 N |
| Quantity | 0.5 g |
| Synonym | Benzen-3,4,5-d3-amine-d2, 2,6-di(ethyl-d5)- (9 CI), 2,6-Di(ethyl-d5)benzen-3,4,5-d3-amine-d2, 2,6-Diethylaniline-d15, 2,6-Diethylaniline-D15, 2,6-Diethylbenzenamine-d15 |
| InChI Formula | InChI=1 S/C10H15N/c1-3-8-6-5-7-9(4-2)10(8)11/h5-7 H,3-4,11H2,1-2H3/i1D3,2D3,3D2,4D2,5 D,6 D,7 D/hD2 |
| SMILES | [2 H]N([2 H])c1c(c([2 H])c([2 H])c([2 H])c1C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N,3,4,5-pentadeuterio-2,6-bis(1,1,2,2,2-pentadeuterioethyl)aniline |
| Molecular Weight (g/mol) | 164.3252 |
| Formula Weight | 164.2146 g/mol |
| Show More |