missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dichloro-3'-fluoroacetophenone, 98%, Thermo Scientific™

Product Code. 11352529
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352529 25 g 25g
11342529 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352529 Supplier Thermo Scientific Alfa Aesar Supplier No. H26638.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 290835-85-7
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.03
MDL Number MFCD02093760
InChI Key VJBFZHHRVCPAPZ-UHFFFAOYSA-N
Synonym 2',6'-dichloro-3'-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethanone, 2,6-dichloro-3-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethan-1-one, 2,6-dichloro-3-fluro acetophenone, 2,6-dichloro-5-fluoro acetophenone, 2',6'-dichloro-3'-fluroacetophenone, ethanone, 1-2,6-dichloro-3-fluorophenyl, 1-acetyl-2,6-dichloro-3-fluorobenzene, pubchem8532
PubChem CID 2733982
IUPAC Name 1-(2,6-dichloro-3-fluorophenyl)ethanone
SMILES CC(=O)C1=C(Cl)C=CC(F)=C1Cl

Specifications

Density 1.403
Boiling Point 255°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 25 g
Formula Weight 207.03
Percent Purity 98%
Chemical Name or Material 2',6'-Dichloro-3'-fluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.